2-Fluorothioacetamide ≥95%

Supplier: Matrix Scientific

115450-1G 115450-5G 115450-500MG
102896-264EA 1033.85 USD
102896-264 102896-266 102896-262
2-Fluorothioacetamide ≥95%
2-Fluorothioacetamide

Hazard: Irritant

Formula: C₂H₄FNS
MDL Number: MFCD19204804
CAS Number: 84350-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR