Amidinothiourea ≥98%
Supplier: Matrix Scientific
Synonyms:
Guanylthiourea
115448-25G
115448-100G
102896-252EA
46.15
USD
102896-252
102896-254
Amidinothiourea ≥98%
Amidinothiourea
Hazard: Irritant
Formula:
C₂H₆N₄S Melting Pt: 170…172 °C |
MDL Number:
MFCD00014472 CAS Number: 2114-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...