1,3-Diphenylparabanic acid ≥98%

Supplier: Matrix Scientific

115445-1G 115445-250MG 115445-500MG
102899-380EA 104.62 USD
102899-380 102896-322 102899-342
1,3-Diphenylparabanic acid ≥98%
1,3-Diphenylparabanic acid

Hazard: Irritant

Formula: C₁₅H₁₀N₂O₃
MDL Number: MFCD00020868
CAS Number: 6488-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR