2,4-Diiodooxazole ≥97%

Supplier: Matrix Scientific

115444-1G 115444-500MG 115444-5G
102899-340EA 3273.85 USD
102899-340 102899-338 102899-336
2,4-Diiodooxazole ≥97%
2,4-Diiodooxazole

Hazard: Irritant

Formula: C₃HI₂NO
MDL Number: MFCD18785731
CAS Number: 1037597-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR