2,2-Difluoro-4-methylenepentanedioic acid ≥97%
Supplier: Matrix Scientific
115443-100MG
115443-250MG
115443-500MG
102899-334EA
2229.23
USD
102899-334
102899-332
102899-330
2,2-Difluoro-4-methylenepentanedioic acid ≥97%
2,2-Difluoro-4-methylenepentanedioic acid
Hazard: Irritant
Formula:
C₆H₆F₂O₄ |
MDL Number:
MFCD19443976 CAS Number: 1307857-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...