(2S,4S)-2-Methylpiperidin-4-ol ≥95%

Supplier: Matrix Scientific

115437-1G
102896-190EA 3900 USD
102896-190
(2S,4S)-2-Methylpiperidin-4-ol ≥95%
(2S,4S)-2-Methylpiperidin-4-ol

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD19219960
CAS Number: 89451-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR