tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate
115435-250MG
115435-1G
102899-314EA
989.23
USD
102899-314
102896-184
tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₆F₃NO₃ MW: 255.24 g/mol |
MDL Number:
MFCD16990772 CAS Number: 644970-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...