tert-Butyl-5-(ethoxy(hydroxy)methyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115418-500MG
102896-064EA
4910.77
USD
102896-064
tert-Butyl-5-(ethoxy(hydroxy)methyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
tert-Butyl-5-(ethoxy(hydroxy)methyl)-3,3-difluoropiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₂₃F₂NO₄ MW: 295.33 g/mol |
MDL Number:
MFCD22370033 CAS Number: 1379811-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...