tert-Butyl-3-(acetoxymethyl)-4-hydroxypiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115414-5G
115414-1G
115414-500MG
102896-038EA
1044.62
USD
102896-038
102896-040
102896-042
tert-Butyl-3-(acetoxymethyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl-3-(acetoxymethyl)-4-hydroxypiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₂₃NO₅ MW: 273.33 g/mol |
MDL Number:
MFCD22199508 CAS Number: 1373502-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...