tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1,2,3-oxathiazinane-3-carboxylate 2,2-dioxide

115405-5G 115405-1G 115405-500MG
102895-956EA 1421.54 USD
102895-956 102895-954 102895-958
tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate ≥95%
tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₅NO₅S
MW: 237.28 g/mol
MDL Number: MFCD22123271
CAS Number: 521267-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR