4-Azaindole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1H-Pyrrolo[3,2-b]pyridine-3-carboxylic acid
115404-1G
115404-250MG
115404-500MG
102896-246EA
1335.38
USD
102896-246
102896-242
102896-244
4-Azaindole-3-carboxylic acid ≥95%
4-Azaindole-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₂ MW: 162.1454 g/mol |
MDL Number:
MFCD11559014 CAS Number: 860496-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...