Methyl-5-aminopyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

115393-500MG 115393-1G
102899-348EA 2326.15 USD
102899-348 102899-346
Methyl-5-aminopyrimidine-2-carboxylate ≥95%
Methyl-5-aminopyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₆H₇N₃O₂
MDL Number: MFCD08234876
CAS Number: 73418-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR