Ethyl-2-(5-fluoropyridin-3-yl)acetate ≥95%
Supplier: Matrix Scientific
115389-1G
102895-936EA
1680
USD
102895-936
Ethyl-2-(5-fluoropyridin-3-yl)acetate ≥95%
Ethyl-2-(5-fluoropyridin-3-yl)acetate
Hazard: Irritant
Formula:
C₉H₁₀FNO₂ MW: 183.18 g/mol |
MDL Number:
MFCD16748979 CAS Number: 39891-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...