7-Boc-2-chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine ≥95%
Supplier: Matrix Scientific
115366-500MG
115366-250MG
115366-1G
102895-798EA
3004.62
USD
102895-798
102895-796
102895-794
7-Boc-2-chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine ≥95%
7-Boc-2-chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine
Hazard: Irritant
Formula:
C₁₂H₁₆ClN₃O₂ MW: 269.73 g/mol |
MDL Number:
MFCD13189593 CAS Number: 1196156-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...