4-(Aminomethyl)pyrrolidin-2-one ≥95%

Supplier: Matrix Scientific

115358-250MG 115358-500MG 115358-1G
102896-102EA 1421.54 USD
102896-102 102896-100 102895-708
4-(Aminomethyl)pyrrolidin-2-one ≥95%
4-(Aminomethyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₅H₁₀N₂O
MDL Number: MFCD09744146
CAS Number: 676627-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR