6,7-Dihydro-5(4H)-cyclopenta[1,3]dioxin-5-one ≥95%
Supplier: Matrix Scientific
115354-500MG
115354-250MG
115354-100MG
102899-280EA
969.23
USD
102899-280
102899-278
102899-276
6,7-Dihydro-5(4H)-cyclopenta[1,3]dioxin-5-one ≥95%
6,7-Dihydro-5(4H)-cyclopenta[1,3]dioxin-5-one
Hazard: Irritant
Formula:
C₇H₈O₃ |
MDL Number:
MFCD00075482 CAS Number: 102306-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...