Furo[2,3-d]pyrimidin-4-amine ≥96%

Supplier: Matrix Scientific

115352-5G 115352-1G
102899-270EA 5893.85 USD
102899-270 102899-268
Furo[2,3-d]pyrimidin-4-amine ≥96%
Furo[2,3-d]pyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₅N₃O
MDL Number: MFCD09834959
CAS Number: 186454-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR