Tributyl(phenylethenyl)tin 90-95%

Supplier: Matrix Scientific

115344-25G 115344-1G 115344-5G
102895-698EA 3586.15 USD
102895-698 102895-696 102895-694
Tributyl(phenylethenyl)tin 90-95%
Tributyl(phenylethenyl)tin

Hazard: Irritant

Formula: C₂₀H₃₄Sn
MDL Number: MFCD01114691
CAS Number: 79159-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR