(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Tetrahydro-3H-pyrrolo[1,2-c][1,2,3]oxathiazole 1,1-dioxide

115340-1G 115340-5G 115340-500MG
102895-678EA 3926.15 USD
102895-678 102895-578 102895-580
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide ≥95%
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide

Hazard: Irritant

Formula: C₅H₉NO₃S
MW: 163.2 g/mol
MDL Number: MFCD22123262
CAS Number: 132635-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR