5-Iodo-2,3-dihydropyridazin-3-one ≥95%
Supplier: Matrix Scientific
115336-1G
115336-5G
102899-256EA
540
USD
102899-256
102899-254
5-Iodo-2,3-dihydropyridazin-3-one ≥95%
5-Iodo-2,3-dihydropyridazin-3-one
Hazard: Irritant
Formula:
C₄H₃IN₂O MW: 221.9800 g/mol |
MDL Number:
MFCD08693620 CAS Number: 825633-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...