Di-tert-butylisobutylsilyl triflate ≥98%

Supplier: Matrix Scientific

115332-5G
102899-232EA 592.31 USD
102899-232
Di-tert-butylisobutylsilyl triflate ≥98%
Di-tert-butylisobutylsilyl triflate

Hazard: Irritant

Formula: C₁₃H₂₇F₃O₃SSi
MDL Number: MFCD20486183
CAS Number: 1314639-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR