4-Bromobenzaldehyde tosylhydrazone ≥98%

Supplier: Matrix Scientific

115331-25G
102899-132EA 569.23 USD
102899-132
4-Bromobenzaldehyde tosylhydrazone ≥98%
4-Bromobenzaldehyde tosylhydrazone

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂O₂S
MDL Number: MFCD01001172
CAS Number: 19350-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR