Ethyl-1-boc-3-cyanoazetidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

115327-1G
102895-554EA 1813.85 USD
102895-554
Ethyl-1-boc-3-cyanoazetidine-3-carboxylate ≥95%
Ethyl-1-boc-3-cyanoazetidine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₄
MDL Number: MFCD17016129
CAS Number: 1105663-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR