endo-7-Amino-9-boc-3-oxa-9-aza-bicyclo[3.3.1]nonane ≥95%

Supplier: Matrix Scientific

115326-1G
102899-128EA 3873.85 USD
102899-128
endo-7-Amino-9-boc-3-oxa-9-aza-bicyclo[3.3.1]nonane ≥95%
endo-7-Amino-9-boc-3-oxa-9-aza-bicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₃
CAS Number: 280762-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR