tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115321-100MG
115321-500MG
115321-250MG
102899-226EA
4555.38
USD
102899-226
102899-224
102899-222
tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₁H₂₀F₂N₂O₂ MW: 250.29 g/mol |
MDL Number:
MFCD18909808 CAS Number: 1303973-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...