6-Benzyl-8-oxa-2,6-diazaspiro[4.5]decan-7-one ≥95%

Supplier: Matrix Scientific

115318-500MG
102895-998EA 3790.77 USD
102895-998
6-Benzyl-8-oxa-2,6-diazaspiro[4.5]decan-7-one ≥95%
6-Benzyl-8-oxa-2,6-diazaspiro[4.5]decan-7-one

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₂
MW: 246.31 g/mol
MDL Number: MFCD22415222
CAS Number: 1389264-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR