Benzyl-3,5-dioxopiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115317-250MG 115317-500MG 115317-1G
102895-070EA 1241.54 USD
102895-070 102895-996 102895-068
Benzyl-3,5-dioxopiperidine-1-carboxylate ≥95%
Benzyl-3,5-dioxopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₄
MW: 247.25 g/mol
MDL Number: MFCD16618536
CAS Number: 1228631-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR