Benzyl-1,6-diazaspiro[3.5]nonane-6-carboxylate ≥95%
Supplier: Matrix Scientific
115315-250MG
115315-500MG
102895-060EA
2116.92
USD
102895-060
102895-062
Benzyl-1,6-diazaspiro[3.5]nonane-6-carboxylate ≥95%
Benzyl-1,6-diazaspiro[3.5]nonane-6-carboxylate
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₂ MW: 260.34 g/mol |
MDL Number:
MFCD20261437 CAS Number: 1334499-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...