Benzyl-2,6-diazaspiro[3.3]heptane-2-carboxylate ≥95%
Supplier: Matrix Scientific
115314-5G
115314-500MG
115314-1G
102895-058EA
6493.85
USD
102895-058
102895-056
102895-054
Benzyl-2,6-diazaspiro[3.3]heptane-2-carboxylate ≥95%
Benzyl-2,6-diazaspiro[3.3]heptane-2-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₆N₂O₂ MW: 232.28 g/mol |
MDL Number:
MFCD17079020 CAS Number: 1211517-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...