Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥97%
Supplier: Matrix Scientific
115312-1G
115312-5G
102895-048EA
516.92
USD
102895-048
102895-046
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥97%
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate
Purity: 97%
CAS: 188944-77-6
MDL Number: MFCD29052155
Molecular Formula: C10H20F6INP-
Molecular Weight: 395,17
Hazard: Irritant
Formula:
C₁₆H₂₂BrF₆N₂P |
CAS Number:
188944-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...