(R)-Pyrrolidin-3-ylmethanol hydrochloride ≥95%
Supplier: Matrix Scientific
115303-1G
115303-500MG
102894-890EA
1680
USD
102894-890
102894-888
(R)-Pyrrolidin-3-ylmethanol hydrochloride ≥95%
(R)-Pyrrolidin-3-ylmethanol hydrochloride
Hazard: Irritant
Formula:
C₅H₁₁NO |
MDL Number:
MFCD09800470 CAS Number: 1227157-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...