1-Methylpiperazin-2-one hydrochloride ≥98%
Supplier: Matrix Scientific
115301-5G
115301-1G
102894-880EA
1593.85
USD
102894-880
102894-882
1-Methylpiperazin-2-one hydrochloride ≥98%
1-Methylpiperazin-2-one hydrochloride
Hazard: Irritant
Formula:
C₅H₁₁ClN₂O |
MDL Number:
MFCD06662557 CAS Number: 109384-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...