2-Amino-4-methylthiophene hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Methylthiophen-2-amine hydrochloride
115290-1G
115290-250MG
115290-500MG
102895-894EA
1526.15
USD
102895-894
102895-892
102899-178
2-Amino-4-methylthiophene hydrochloride ≥95%
2-Amino-4-methylthiophene hydrochloride
Hazard: Irritant
Formula:
C₅H₈ClNS MW: 149.64 g/mol |
MDL Number:
MFCD21604144 CAS Number: 930299-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...