cis-3-(Aminomethyl)cyclobutanol hydrochloride ≥95%

Supplier: Matrix Scientific

115281-500MG 115281-1G
102895-762EA 1044.62 USD
102895-762 102891-290
cis-3-(Aminomethyl)cyclobutanol hydrochloride ≥95%
cis-3-(Aminomethyl)cyclobutanol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD22421655
CAS Number: 1400744-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR