5-Bromo-3-methylimidazo[1,2-a]pyridine hydrobromide ≥97%
Supplier: Matrix Scientific
115279-5G
115279-500MG
115279-1G
102895-756EA
5793.85
USD
102895-756
102894-630
102894-632
5-Bromo-3-methylimidazo[1,2-a]pyridine hydrobromide ≥97%
5-Bromo-3-methylimidazo[1,2-a]pyridine hydrobromide
Hazard: Irritant
Formula:
C₈H₈Br₂N₂ MW: 291.98 g/mol |
MDL Number:
MFCD22422284 CAS Number: 74420-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...