(5-Chloro-2-pyridyl)methanamine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)-5-chloropyridine dihydrochloride

115276-1G
102894-624EA 307.69 USD
102894-624
(5-Chloro-2-pyridyl)methanamine dihydrochloride ≥95%
(5-Chloro-2-pyridyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₆H₉Cl₃N₂
MW: 215.51 g/mol
MDL Number: MFCD16872047
CAS Number: 1257535-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR