SS-miniPHOS-diborane ≥95%

Supplier: Matrix Scientific

115265-500MG
102895-748EA 515.38 USD
102895-748
SS-miniPHOS-diborane ≥95%
SS-miniPHOS-diborane

Hazard: Irritant

Formula: C₁₁H₃₂B₂P₂
MDL Number: MFCD28099793
CAS Number: 265298-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR