(S,S)-1,2-Bis[(2-methylphenyl)(phenyl)phosphino]ethane ≥95%

Supplier: Matrix Scientific

115243-500MG 115243-1G
102893-038EA 2061.54 USD
102893-038 102893-036
(S,S)-1,2-Bis[(2-methylphenyl)(phenyl)phosphino]ethane ≥95%
(S,S)-1,2-Bis[(2-methylphenyl)(phenyl)phosphino]ethane

Hazard: Irritant

Formula: C₂₈H₂₈P₂
Melting Pt: 86…90 °C
MDL Number: MFCD12198306
CAS Number: 810667-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR