(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥95%
Supplier: Matrix Scientific
Synonyms:
(R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine]
, (R,R)-DIPAMP
, (R,R)-(-)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane
, (1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
, (-)-DIPAMP
115240-1G
115240-500MG
102893-026EA
1238.46
USD
102893-026
102893-024
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥95%
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Hazard: Irritant
Formula:
C₂₈H₂₈O₂P₂ MW: 458.48 g/mol Melting Pt: 106…110 °C |
MDL Number:
MFCD05863546 CAS Number: 55739-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...