(4-Bromophenyl)methanesulfonamide ≥97%

Supplier: Matrix Scientific

115235-1G 115235-500MG
102895-668EA 564.62 USD
102895-668 102895-666
(4-Bromophenyl)methanesulfonamide ≥97%
(4-Bromophenyl)methanesulfonamide

Hazard: Irritant

Formula: C₇H₈BrNO₂S
MW: 250.12 g/mol
MDL Number: MFCD12776301
CAS Number: 64732-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR