4-Methoxy-2,6-dimethylbenzaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
2,6-Dimethyl-4-methoxybenzaldehyde
115230-500MG
115230-1G
102895-652EA
843.08
USD
102895-652
102895-650
4-Methoxy-2,6-dimethylbenzaldehyde ≥97%
4-Methoxy-2,6-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₂O₂ MW: 164.21 g/mol |
MDL Number:
MFCD04971199 CAS Number: 19447-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...