4-Formyl-3,5-dimethylphenyl acetate ≥97%
Supplier: Matrix Scientific
115227-500MG
115227-1G
102895-638EA
624.62
USD
102895-638
102895-640
4-Formyl-3,5-dimethylphenyl acetate ≥97%
4-Formyl-3,5-dimethylphenyl acetate
Hazard: Irritant
Formula:
C₁₁H₁₂O₃ Melting Pt: 53…55 °C |
MDL Number:
MFCD24686316 CAS Number: 925441-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...