2-Hydroxy-4-methanesulfonylbenzoic acid ≥97%
Supplier: Matrix Scientific
115225-1G
115225-500MG
102892-578EA
1453.85
USD
102892-578
102892-580
2-Hydroxy-4-methanesulfonylbenzoic acid ≥97%
2-Hydroxy-4-methanesulfonylbenzoic acid
Hazard: Irritant
Formula:
C₈H₈O₅S MW: 216.21 g/mol |
MDL Number:
MFCD24483010 CAS Number: 108153-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...