(5-Fluoro-1-benzofuran-2-yl)methanol ≥97%

Supplier: Matrix Scientific

115216-500MG
102899-142EA 1453.85 USD
102899-142
(5-Fluoro-1-benzofuran-2-yl)methanol ≥97%
(5-Fluoro-1-benzofuran-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇FO₂
MDL Number: MFCD09907889
CAS Number: 276235-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR