Methyl-2-amino-3-hydroxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-3-hydroxybenzoate

115215-1G
102895-612EA 361.54 USD
102895-612
Methyl-2-amino-3-hydroxybenzoate ≥97%
Methyl-2-amino-3-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD01675233
CAS Number: 17672-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR