2-Bromo-6-chlorophenol ≥97%
Supplier: Matrix Scientific
115208-1G
102892-406EA
264.62
USD
102892-406
2-Bromo-6-chlorophenol ≥97%
2-Bromo-6-chlorophenol
Hazard: Irritant
Formula:
C₆H₄BrClO |
MDL Number:
MFCD09907875 CAS Number: 2040-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...