4-Propyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
115181-500MG
115181-1G
102895-536EA
1040
USD
102895-536
102895-534
4-Propyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid ≥95%
4-Propyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₂S |
MDL Number:
MFCD14797777 CAS Number: 1031672-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...