2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

115165-500MG 115165-1G
102892-190EA 693.85 USD
102892-190 102892-192
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid ≥95%
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₃
MDL Number: MFCD20754075
CAS Number: 1111000-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR