N-(3-Acetylphenyl)-2-azidoacetamide ≥95%

Supplier: Matrix Scientific

115149-500MG 115149-1G
102891-894EA 1040 USD
102891-894 102891-844
N-(3-Acetylphenyl)-2-azidoacetamide ≥95%
N-(3-Acetylphenyl)-2-azidoacetamide

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₂
MDL Number: MFCD14652701
CAS Number: 1248994-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR