3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride ≥95%

Supplier: Matrix Scientific

115145-1G 115145-500MG
102899-094EA 1040 USD
102899-094 102899-092
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride ≥95%
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂Cl₂N₄
MDL Number: MFCD25975127
CAS Number: 1443423-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR